Learn More
IL-38 Monoclonal Antibody (H127C), Biotin, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 13-7385-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The monoclonal antibody H127C recognizes human IL-38. IL-38 is a member of the Interleukin-1 family (IL-1F) and shares homology with IL-1R antagonist (IL-1Ra) and IL-36R antagonist (IL-36Ra). IL-38 is expressed mostly in the skin and proliferating B cells of the tonsil and is also present at low levels in the heart and placenta. IL-38 functions as an anti-inflammatory cytokine that inhibits IL-36 by binding to IL-36R and also inhibits the production of IL-17 and IL-22. Gene polymorphisms of IL-38 are associated with psoriatic arthritis, ankylosing spondylitis, and heart disease. Applications Reported: This H127C antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This H127C antibody has been tested tested by immunohistochemistry on human FFPE tissue using low pH antigen retrieval and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
Description: The monoclonal antibody H127C recognizes human IL-38. IL-38 is a member of the Interleukin-1 family (IL-1F) and shares homology with IL-1R antagonist (IL-1Ra) and IL-36R antagonist (IL-36Ra). IL-38 is expressed mostly in the skin and proliferating B cells of the tonsil and is also present at low levels in the heart and placenta. IL-38 functions as an anti-inflammatory cytokine that inhibits IL-36 by binding to IL-36R and also inhibits the production of IL-17 and IL-22. Gene polymorphisms of IL-38 are associated with psoriatic arthritis, ankylosing spondylitis, and heart disease.Applications Reported: This H127C antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.Applications Tested: This H127C antibody has been tested tested by immunohistochemistry on human FFPE tissue using low pH antigen retrieval and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.Specifications
IL-38 | |
Monoclonal | |
0.5mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
Q8WWZ1 | |
IL1F10 | |
Affinity chromatography | |
RUO | |
84639 | |
Store at 2-8°C. Do not freeze. | |
Liquid |
Immunohistochemistry (Paraffin), Immunohistochemistry | |
H127C | |
Biotin | |
IL1F10 | |
IL-38, FKSG75, IL1HY2, IL-1HY2, IL1-theta, FIL1-theta, FIL1T, IL38, UNQ6119/PRO20041, Interleukin-1 family member 10, Family of interleukin 1-theta, IL-1F10 | |
Mouse | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
For Research Use Only.