missing translation for 'onlineSavingsMsg'
Learn More

IL-38/IL-1F10 Antibody (798036) [DyLight 755], Novus Biologicals™

Product Code. 18543997 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18543997 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18543997 Supplier Novus Biologicals Supplier No. FAB7774Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-38/IL-1F10 Monoclonal antibody specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-38/IL-1F10
Applications Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 798036
Conjugate DyLight 755
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias interleukin 1 family, member 10 (theta), UNQ6119/PRO20041
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-38/IL-1F10, Met1-Arg152, Accession # Q8R459
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 84639
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.