Learn More
Invitrogen™ IL-37 Monoclonal Antibody (37D12), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 14-7379-37
Description
Description: This 37D12 monoclonal antibody reacts with human IL-37. IL-37 is a member of the interleukin 1 cytokine family and is also known as IL-1F7 and IL-1H4. IL-37 suppresses innate inflammatory and immune responses and exhibits anti-inflammatory characteristics resembling those of IL-1R8. IL-37 binds to the alpha chain of IL-18R with low affinity and does not recruit the essential signaling co-receptor IL-18 beta or mediate pro-inflammatory activity. IL-37 also binds to IL-18 binding protein (IL-18BP) and enhances the antagonistic effects of IL-18BP. IL-37 forms a trimeric complex with IL-18BP and IL-18R beta, which results in the blockade of IL-18 activity. Transcripts of IL-37 have been detected in human tissues, such as lung, testis, and colon tumors, as well as some human cell lines. Applications Reported: This 37D12 antibody has been reported for use in immunocytochemistry followed by cytospin. Applications Tested: This 37D12 antibody has been tested by immunocytochemistry of fixed and permeabilized normal human peripheral blood mononuclear cells and can be used at less than or equal to 20 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). This cytokine also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18. This gene along with eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Five alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| IL-37 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q9NZH6 | |
| IL37 | |
| Affinity chromatography | |
| RUO | |
| 27178 | |
| 4°C | |
| Liquid |
| Immunocytochemistry | |
| 37D12 | |
| Unconjugated | |
| IL37 | |
| FIL1; FIL1 zeta; FIL1(ZETA); FIL1Z; IL-1 zeta; IL1F7; IL-1F7; IL1F7 (canonical product IL-1F7b); IL-1F7b (IL-1H4, IL-1H, IL-1RP1); IL-1H; IL1H4; IL-1H4; IL1RP1; IL-1RP1; IL-1X; IL-1X protein; IL37; IL-37; ILN; Interleukin; interleukin 1 family member 7; interleukin 1, zeta; interleukin 37; Interleukin-1 family member 7; Interleukin-1 homolog 4; interleukin-1 superfamily z; Interleukin-1 zeta; Interleukin-1-related protein; interleukin-23; Interleukin37; Interleukin-37 | |
| Mouse | |
| 2 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.