missing translation for 'onlineSavingsMsg'
Learn More

IL-37/IL-1F7 Antibody (899826) [Janelia Fluor™ 585], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB19751JF585

Product Code. 30167336

  • £452.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IL-37/IL-1F7 Monoclonal antibody specifically detects IL-37/IL-1F7 in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-37/IL-1F7
Monoclonal
Janelia Fluor 585
FIL1, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, interleukin 1 family, member 7 (zeta), interleukin 1, zeta, interleukin-1 family member 7, Interleukin-1 homolog 4, interleukin-1 superfamily z, Interleukin-1 zeta, Interleukin-1-related protein
E. coli-derived recombinant human IL-37/IL-1F7, Lys53-Val206, Accession # Q9NZH6
RUO
Primary
Human
Liquid
Flow Cytometry, CyTOF
899826
50mM Sodium Borate
Mouse
0.1 mL
Cytokine Research
27178
Store at 4°C in the dark.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.