missing translation for 'onlineSavingsMsg'
Learn More

IL-36Ra/IL-1F5 Antibody (022), FITC, Novus Biologicals™

Product Code. 18338260 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338260 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338260 Supplier Novus Biologicals Supplier No. NBP290483F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-36Ra/IL-1F5 Monoclonal specifically detects IL-36Ra/IL-1F5 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-36Ra/IL-1F5
Applications ELISA
Classification Monoclonal
Clone 22
Conjugate FITC
Formulation PBS
Gene Alias FIL1, FIL1 delta, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta), FIL1DIL-1 delta, IL1F5 (Canonical product IL-1F5a), IL-1RP3, IL36RA, IL-36RA, interleukin 1 family, member 5 (delta), interleukin 1, delta, Interleukin 36 Receptor Antagonist, interleukin-1 family member 5, Interleukin-1 HY1, Interleukin-1 receptor antagonist homolog 1, Interleukin-1-like protein 1, Interleukin-36 Receptor Antagonist
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-36Ra/IL-1F5 (NP_062324.2; Val3-Asp156).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 26525
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.