missing translation for 'onlineSavingsMsg'
Learn More

IL-36 gamma/IL-1F9 Antibody (Jussy-1), DyLight 680, Novus Biologicals™

Product Code. 18387412 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387412 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387412 Supplier Novus Biologicals Supplier No. NBP311676FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-36 gamma/IL-1F9 Monoclonal specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-36 gamma/IL-1F9
Applications Western Blot, ELISA
Classification Monoclonal
Clone Jussy-1
Conjugate DyLight 680
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias IL-1 epsilon, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1RP2, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma
Host Species Mouse
Immunogen Recombinant human IL-36 gamma/IL-1F9.
Purification Method >95%, by SDS-PAGE
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 56300
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.