missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-36 gamma/IL-1F9 Antibody (Jussy-1), Alexa Fluor™ 532, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IL-36 gamma/IL-1F9 Monoclonal specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | IL-36 gamma/IL-1F9 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | Jussy-1 |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | IL-1 epsilon, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1RP2, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma |
| Host Species | Mouse |
| Immunogen | Recombinant human IL-36 gamma/IL-1F9. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?