missing translation for 'onlineSavingsMsg'
Learn More

IL-36 Beta/IL-1F8 Mouse anti-Human, CoraLite 594, Clone: 10D12B1, Proteintech

Product Code. 17422653 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17422653 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17422653 Supplier Proteintech Supplier No. CL59460290100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The IL-36 Beta/IL-1F8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-36 Beta/IL-1F8. This antibody recognizes human antigen. The IL-36 Beta/IL-1F8 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.

Specifications

Antigen IL-36 Beta/IL-1F8
Applications Immunofluorescence
Classification Monoclonal
Clone 10D12B1
Conjugate CoraLite 594
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 27177
Target Species Human
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.