missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IL-36 beta/IL-1F8 Antibody Pair

Product Code. 18636271 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18636271

Brand: Novus Biologicals™ NBP278859

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pair

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 10 pg/mL
Conjugate Unconjugated
Product Type Sandwich ELISA
Sample Type Cell Culture Supernatant
For Use With (Application) Sandwich ELISA
Gene ID (Entrez) 27177
Gene Symbol IL36B
Quantity 1 Set
Research Discipline Biologically Active Proteins
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Synonym family of interleukin 1-eta, FIL1, FIL1 eta, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1 eta, IL1-ETA, IL1F8, IL-1F8, IL1F8 (Canonical product IL-1F8a), IL-1F8 (FIL1-eta), IL1H2, IL-1H2, interleukin 1 family, member 8 (eta), interleukin 1, eta, Interleukin 36, Beta, Interleukin-1 eta, interleukin-1 family member 8, Interleukin-1 homolog 2, Interleukin-1 Superfamily e, Interleukin-36 Beta
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.