missing translation for 'onlineSavingsMsg'
Learn More

IL-36 beta/IL-1F8 Antibody (MM0387-2Q27), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18407636 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407636 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407636 Supplier Novus Biologicals Supplier No. NBP211687JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-36 beta/IL-1F8 Monoclonal antibody specifically detects IL-36 beta/IL-1F8 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-36 beta/IL-1F8
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0387-2Q27
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias family of interleukin 1-eta, FIL1, FIL1 eta, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1 eta, IL1-ETA, IL1F8, IL-1F8, IL1F8 (Canonical product IL-1F8a), IL-1F8 (FIL1-eta), IL1H2, IL-1H2, interleukin 1 family, member 8 (eta), interleukin 1, eta, Interleukin 36, Beta, Interleukin-1 eta, interleukin-1 family member 8, Interleukin-1 homolog 2, Interleukin-1 Superfamily e, Interleukin-36 Beta
Host Species Mouse
Immunogen Human recombinant IL-36 beta/IL-1F8
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 27177
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.