missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-36 alpha/IL-1F6 Rabbit anti-Human, Clone: 112, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | IL-36 alpha/IL-1F6 |
|---|---|
| Clone | 112 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18236946
|
Novus Biologicals
NBP2-89583-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18217457
|
Novus Biologicals
NBP2-89583-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-36 alpha/IL-1F6 Monoclonal specifically detects IL-36 alpha/IL-1F6 in Human samples. It is validated for ELISA.Specifications
| IL-36 alpha/IL-1F6 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha | |
| Recombinant Human IL-36 alpha/IL-1F6 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 112 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins | |
| 0.2 um filtered solution in PBS | |
| 27179 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title