missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-33R (ST2) Recombinant Rabbit Monoclonal Antibody (ST2IL33R-H2), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£301.00 - £600.00

Specifications

Antigen IL-33R (ST2)
Clone ST2IL33R-H2
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17187038
View Documents
Invitrogen™
MA537452
20 μL
£301.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17106798
GREENER_CHOICE
View Documents
Invitrogen™
MA537204
200 μL
£600.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-33R, also known as ST2, is a receptor for IL-33. It is a member of the IL-1 receptor family of innate receptors. Its extracellular domain consists of 3 Ig-like domains that directly interact with its co-receptor IL-1RAcP, and the ligand alarmin, IL-33. IL-33 ligation results in the association of IL-33R complex with its adaptor proteins MyD88, IRAK1, IRAK4 and TRAF6, and activation of the NF-kB, and MAPK pathways. This, in turn, instigates the release of chemokines and cytokines such as IL-5, IL-6, IL-8, and IL-13. Two most common isoforms of IL-33R that result from alternative splicing are ST2L – the membrane bound form, and ST2S – the soluble form. The membrane bound form can be released from the cell surface by proteolytic cleavage. The soluble forms have been suggested to act as decoys dampening the IL-33 signaling. IL-33R is primarily expressed by mast cells, basophils, and eosinophils. In mouse, IL-33R expression has been shown on ILC2 and Th2 cells. IL-33R signaling has been implicated in the number of immune conditions, including allergies, arthritis, atherosclerosis and sepsis.
Specifications

Specifications

IL-33R (ST2)
0.5 mg/mL
Recombinant Monoclonal
Liquid
RUO
PBS with 0.20% BSA and 0.09% sodium azide
DER4; Fit1; Fit-1; Fos-responsive gene 1 protein; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; IL-1 R4; IL1RL1; IL-1RL1; IL33R; IL-33R; interleukin 1 receptor like 1; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin1 receptor like 1; interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Ly84; lymphocyte antigen 84; Protein ST2; Protein T1; sIL 33R; soluble IL 33 Receptor; soluble IL 33R; St2; ST2L; St2-rs1; ST2V; Ste2; T1; T1/ST2
IL1RL1
IgG κ
Protein A/G
Antibody
ST2IL33R-H2
Flow Cytometry
Unconjugated
Rabbit
Human
Q01638
9173
human recombinant ST2-Fc.
Primary
-20°C
IL1RL1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.