missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-33R (ST2) Monoclonal Antibody (RMST2-2), Brilliant Ultra Violet™ 615, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£183.00 - £369.00
Specifications
| Antigen | IL-33R (ST2) |
|---|---|
| Clone | RMST2-2 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17972161
|
Invitrogen™
366-9335-80 |
25 μg |
£183.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17932091
![]() |
Invitrogen™
366-9335-82 |
100 μg |
£369.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-33R, also known as ST2, is a receptor for IL-33. It is a member of the IL-1 receptor family of innate receptors. Its extracellular domain consists of 3 Ig-like domains that directly interact with its co-receptor IL-1RAcP, and the ligand alarmin, IL-33. IL-33 ligation results in the association of IL-33R complex with its adaptor proteins MyD88, IRAK1, IRAK4 and TRAF6, and activation of the NF-kB, and MAPK pathways. This, in turn, instigates the release of chemokines and cytokines such as IL-5, IL-6, IL-8, and IL-13. Two most common isoforms of IL-33R that result from alternative splicing are ST2L – the membrane bound form, and ST2S – the soluble form. The membrane bound form can be released from the cell surface by proteolytic cleavage. The soluble forms have been suggested to act as decoys dampening the IL-33 signaling. IL-33R is primarily expressed by mast cells, basophils, and eosinophils. In mouse, IL-33R expression has been shown on ILC2 and Th2 cells. IL-33R signaling has been implicated in the number of immune conditions, including allergies, arthritis, atherosclerosis and sepsis.Specifications
| IL-33R (ST2) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| DER4; Fit1; Fit-1; Fos-responsive gene 1 protein; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; IL-1 R4; IL1RL1; IL-1RL1; IL33R; IL-33R; interleukin 1 receptor like 1; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin1 receptor like 1; interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Ly84; lymphocyte antigen 84; Protein ST2; Protein T1; sIL 33R; soluble IL 33 Receptor; soluble IL 33R; St2; ST2L; St2-rs1; ST2V; Ste2; T1; T1/ST2 | |
| IL1RL1 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| RMST2-2 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 615 | |
| Rat | |
| Mouse | |
| P14719 | |
| 17082 | |
| IgG2a κ | |
| Affinity chromatography | |
| IL1RL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title