Learn More
Invitrogen™ IL-33 Polyclonal Antibody

Description
The PA5-20398 immunogen is located within the 150-200 amino acids of IL-33. Predicted molecular ~ 31kD. In Western blot applications, this antibody has been observed to detect a band at: 33kD. Despite its predicted molecular weight, IL-33 often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is human lymph node tissue lysate. PA5-20398 can be used with blocking peptide PEP-0515. Predicted species reactivity based on immunogen sequence: Rat: (76.5%).
Specifications
Specifications
| Antigen | IL-33 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | IL33 |
| Gene Accession No. | O95760, P14719, Q01638, Q8BVZ5 |
| Gene Alias | 9230117N10Rik; C9orf26; DVS27; DVS27-related protein; H-IL-33; IL1F11; IL-1F11; Il33; IL-33; ILN; Interleukin; interleukin 33; interleukin-1 family member 11; Interleukin33; Interleukin-33; Interleukin-33 (109-270); Interleukin-33 (95-270); Interleukin-33 (99-270); Interleukin-33(102-266); Interleukin-33(109-266); NFEHEV; NFHEV; NF-HEV; nuclear factor for high endothelial venules; nuclear factor from high endothelial venules; RGD1311155; RP11-575C20.2 |
| Gene Symbols | IL1rl1, IL33 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.