missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-32 Recombinant Rabbit Monoclonal Antibody (403), HRP

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552279
This item is not returnable.
View return policy
Description
IL-32 Recombinant Monoclonal Antibody for ELISA
Interleukin-32 (IL-32) was initially identified as a transcript (NK4) that is selectively expressed in lymphocytes and NK cells and whose expression is increased following activation by IL-2. It was later re-isolated from an IL-18-treated lung carcinoma cell line and re-named IL-32. IL-32 is unusual in that it does not share sequence homology with known cytokine families and is highly expressed in immune tissues, existing in at least four differentially spliced isoforms. Because treatment of human monocytic and mouse macrophage cells with IL-32 induces several proinflammatory cytokines such as TNF-a, IL-8 and MIP-2, and because it is also induced in human peripheral lymphocyte cells after mitogen stimulation and in epithelial cells by IFN-gamma, it has been suggested that IL-32 may play a role in autoimmune and inflammatory diseases such as rheumatoid arthritis.
Specifications
| IL-32 | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with no preservative | |
| P24001 | |
| IL32 | |
| Recombinant Human IL32 Protein (Met1-Lys131). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 403 | |
| HRP | |
| IL32 | |
| IL32; IL-32; IL-32alpha; IL-32beta; IL-32delta; IL-32gamma; ILN; Interleukin; interleukin 32; interleukin-32; interleukin-32 eta; interleukin-32 small; interleukin-32 theta; natural killer cell transcript 4; natural killer cells protein 4; NK4; TAIF; TAIFa; TAIFb; TAIFc; TAIFd; Tumor necrosis factor alpha-inducing factor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 9235 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu