missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-32 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA575449
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
IL-32 is a 234 amino acid member of the cytokine family which plays an important role in innate and adaptive immune responses. It contains a tyrosine sulfation site, three potential N-myristoylation sites, multiple putative phosphorylation sites and a RGD cell-attachment sequence. IL-32 shows increased expression after activation of T lymphocytes and natural killer cells by IL-2 or mitogens. Human IL-32 exists as four splice variants. Reports suggest that it activates typical cytokine signal pathways of nuclear factor-kappa B (NF-kappaB) and p38 mitogen-activated protein kinase. It induces the production of various cytokines such as TNFA/TNF-alpha, macrophage inflammatory protein (MIP)-2 and IL-8, and has a possible role in inflammatory/autoimmune diseases like chronic obstructive pulmonary disease, Crohn′s disease and psoriasis. It is ubiquitously expressed in most tissues with selective expression in lymphocytes and more prominently in immune cells.
Specifications
| IL-32 | |
| Polyclonal | |
| Unconjugated | |
| IL32 | |
| Interleukin-32; IL-32; Natural killer cells protein 4; Tumor necrosis factor alpha-inducing factor; NK4; TAIF; IL32 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9235 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P24001 | |
| IL32 | |
| Synthetic peptide corresponding to amino acids 200-2450 of Human IL-32. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction