missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-32 Monoclonal Antibody (373802)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561249

Product Code. 30534593

  • £315.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

Interleukin-32 (IL-32) was initially identified as a transcript (NK4) that is selectively expressed in lymphocytes and NK cells and whose expression is increased following activation by IL-2. It was later re-isolated from an IL-18-treated lung carcinoma cell line and re-named IL-32. IL-32 is unusual in that it does not share sequence homology with known cytokine families and is highly expressed in immune tissues, existing in at least four differentially spliced isoforms. Because treatment of human monocytic and mouse macrophage cells with IL-32 induces several proinflammatory cytokines such as TNF-a, IL-8 and MIP-2, and because it is also induced in human peripheral lymphocyte cells after mitogen stimulation and in epithelial cells by IFN-gamma, it has been suggested that IL-32 may play a role in autoimmune and inflammatory diseases such as rheumatoid arthritis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-32
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P24001
IL32
E. coli-derived recombinant human IL-32 alpha , Phe3-Lys131.
100 μg
Primary
Human
Antibody
IgG2a
Western Blot, Immunocytochemistry
373802
Unconjugated
IL32
IL32; IL-32; IL-32alpha; IL-32beta; IL-32delta; IL-32gamma; ILN; Interleukin; interleukin 32; interleukin-32; interleukin-32 eta; interleukin-32 small; interleukin-32 theta; natural killer cell transcript 4; natural killer cells protein 4; NK4; TAIF; TAIFa; TAIFb; TAIFc; TAIFd; Tumor necrosis factor alpha-inducing factor
Rat
Protein A/G
RUO
9235
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.