missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-32 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £238.00
Specifications
| Antigen | IL-32 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620392
|
Novus Biologicals
NBP2-95285-100ul |
100 μg | |||||||
|
18661624
|
Novus Biologicals
NBP2-95285-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-32 Polyclonal antibody specifically detects IL-32 in Human samples. It is validated for Western BlotSpecifications
| IL-32 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 9235 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4IL-32delta, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human IL-32 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title