missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-32 Polyclonal specifically detects IL-32 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | IL-32 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4IL-32delta, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant IL-32 (NP_001012651.1; Met1-Lys131). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?