missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-32 alpha Antibody (12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06577-100ul
This item is not returnable.
View return policy
Description
IL-32 alpha Monoclonal antibody specifically detects IL-32 alpha in Human samples. It is validated for Western Blot
Specifications
| IL-32 alpha | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 9235 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot | |
| 12 | |
| Western Blot 1:500-1:1000 | |
| IL32, IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, interleukin 32, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-32 alpha (Accession#: NP_001012651.1; Met1-Lys131). | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction