Learn More
IL-32 alpha Antibody (12), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06577H
Product Code. 18605498
  • £329.00 / 0.10mL
Description
IL-32 alpha Monoclonal antibody specifically detects IL-32 alpha in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-32 alpha
Monoclonal
HRP
PBS
Mouse
Protein A purified
RUO
9235
Store at 4°C in the dark.
IgG
Western Blot
12
Western Blot
IL32, IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, interleukin 32, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-32 alpha (NP_001012651.1; Met1-Lys131).
0.1 mL
Primary
Human
Purified