missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-31RA Antibody (313308), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2769CL1
This item is not returnable.
View return policy
Description
IL-31RA Monoclonal antibody specifically detects IL-31RA in Human samples. It is validated for Flow Cytometry
Specifications
| IL-31RA | |
| Monoclonal | |
| CoraFluor 1 | |
| class I cytokine receptor, CRL3Glmr, Cytokine receptor-like 3, GLMR, GLM-RMGC125346, GP130 like receptor, Gp130-like monocyte receptor, Gp130-like receptor, hGLM-R, IL-31 receptor subunit alpha, IL-31R subunit alpha, IL-31RAGPLCRL, IL-31R-alpha, interleukin 31 receptor A, interleukin-31 receptor subunit alpha, PRO21384, soluble type I cytokine receptor CRL3, ZcytoR17 | |
| NS0-derived rhIL-31 RA extracellular domain, aa 20-516, Accession # NP_620586 | |
| 0.1 mL | |
| Apoptosis | |
| 133396 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 313308 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction