missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-31 Polyclonal Antibody
Description
Despite its predicted molecular weight, IL-31 often migrates at ~27-30kDa in SDS-PAGE. A suggested positive control is human skeletal muscle tissue lysate. PA5-20220 can be used with blocking peptide PEP-0333.
Interleukin-31 (IL-31) is a recently discovered T-cell cytokine closely related to IL-6 type cytokines and is preferentially produced by T helper type 2 cells. IL-31 activity is mediated through the ligand-induced oligomerization of a dimeric receptor complex containing IL-31 receptor A and oncostatin M receptor. In response to IL-31 binding, these proteins activate the JAK/STAT and the AKT signaling pathways. RNA levels of IL-31 receptor A and oncostatin M receptor are induced in activated monocytes but are expressed constitutively in epithelial cells. IL-31, when overexpressed in transgenic mice, results in the development of pruritis, alopecia, and skin lesions and in humans may result in atopic dermatitis, suggesting that IL-31 may represent a novel target for antipruritic drug development.
Specifications
Specifications
| Antigen | IL-31 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Il31 |
| Gene Accession No. | Q6EBC2 |
| Gene Alias | 1700013B14Rik; IL31; IL-31; ILN; Interleukin; interleukin 31; interleukin-31 |
| Gene Symbols | Il31 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?