missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526394
This item is not returnable.
View return policy
Description
IL-31 Polyclonal Antibody for Western Blot, IHC (P), Flow
Interleukin-31 (IL-31) is a recently discovered T-cell cytokine closely related to IL-6 type cytokines and is preferentially produced by T helper type 2 cells. IL-31 activity is mediated through the ligand-induced oligomerization of a dimeric receptor complex containing IL-31 receptor A and oncostatin M receptor. In response to IL-31 binding, these proteins activate the JAK/STAT and the AKT signaling pathways. RNA levels of IL-31 receptor A and oncostatin M receptor are induced in activated monocytes but are expressed constitutively in epithelial cells. IL-31, when overexpressed in transgenic mice, results in the development of pruritis, alopecia, and skin lesions and in humans may result in atopic dermatitis, suggesting that IL-31 may represent a novel target for antipruritic drug development.
Specifications
| IL-31 | |
| Polyclonal | |
| Unconjugated | |
| Il31 | |
| 1700013B14Rik; IL31; IL-31; ILN; Interleukin; interleukin 31; interleukin-31 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 386653 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6EBC2 | |
| Il31 | |
| KLH conjugated synthetic peptide between 13-40 amino acids from the N-terminal region of human IL31. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction