missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-31 Mouse ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ BMS6030

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15552007

  • £451.00 / 96 tests
Estimated Shipment: 22-05-2024
to see stock.
Add to basket

Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-31
Biotin-Conjugate anti-mouse IL-31 monoclonal antibody
Streptavidin-HRP
Mouse IL-31 Standard lyophilized, 2000pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

Description

Description

The Mouse Interleukin-31 (Ms IL-31) ELISA quantitates Ms IL-31 in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-31. Principle of the method The Mouse IL-31 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Interleukin 31, or IL-31 is a cytokine secreted by differentiated CD4+ T helper type 2 (Th2) cells. It is also released during inflammation of the skin, by macrophages and T cells. Because IL-31 is secreted by activated Th2 cells, it has a role in host defense against infection and may be involved in asthma, allergy and different types of bowel diseases.
Specifications

Specifications

Q6EAL8
9.1 pg/mL
ELISA Kit
Mouse
Colorimetric Microplate Reader
1700013B14Rik
7.2%
HRP
RUO
2°C to 8°C
Mouse
3 hr. 30 min.
15.63-1,000 pg/mL
Biotin
Plasma, Serum, Supernatant
ELISA
76399
5.6%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
Interleukin-31
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers