missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-31 ELISA Kit

Product Code. 17649306
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17649306

Brand: Invitrogen™ BMS20412

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Human Interleukin-31 (Hu IL-31) ELISA quantitates Hu IL-31 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-31. Principle of the method The Human IL-31 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Interleukin-31 (IL-31) is a recently discovered T-cell cytokine closely related to IL-6 type cytokines and is preferentially produced by T helper type 2 cells. IL-31 activity is mediated through the ligand-induced oligomerization of a dimeric receptor complex containing IL-31 receptor A and oncostatin M receptor. In response to IL-31 binding, these proteins activate the JAK/STAT and the AKT signaling pathways. RNA levels of IL-31 receptor A and oncostatin M receptor are induced in activated monocytes but are expressed constitutively in epithelial cells. IL-31, when overexpressed in transgenic mice, results in the development of pruritis, alopecia, and skin lesions and in humans may result in atopic dermatitis, suggesting that IL-31 may represent a novel target for antipruritic drug development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6EBC2
Assay Range 7.8-500 pg/mL
Assay Sensitivity 1.2 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1700013B14Rik; IL31; IL-31; ILN; Interleukin; interleukin 31; interleukin-311700013B14Rik; IL31; IL-31; ILN; Interleukin; interleukin 31; interleukin-31;
Gene ID (Entrez) 386653
Gene Symbol Il31
Interassay CV 3.3%
Intraassay CV 5.7%
Kit Contents Pre-coated 96 well plate, Standard (2 vials), Sample diluent, Biotinylated Detection Antibody, Assay buffer Concentrate, Wash buffer concentrate,SAV-HRP,TMB, Stop solution, Adhesive Plate Covers
Label or Dye Biotin
Protein IL-31
Protein Aliases IL-31, Interleukin-31
Quantity 96 Tests
Regulatory Status RUO
Shipping Condition Wet Ice
Storage Requirements 2-8°C
Target IL-31
Target Species Validated Human
Test Time 1 hr 20 min
Total Assay Time 4 hr 30 min
UniProt ID Q6EBC2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.