missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-3 ProQuantum Immunoassay Kit
GREENER_CHOICE

Product Code. 15950473
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
96 reactions
Each
This item is not returnable. View return policy

Product Code. 15950473

Brand: Invitrogen™ A35586

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08700
Assay Range 0.0128 to 5000 pg/mL
Assay Sensitivity <0.01 pg/mL
Conjugate FAM™
Detection Method Taqman™ Real-time PCR
Immunoassay Kit Format Proximity-based Immunoassay
Product Type ProQuantum
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) Protein Quantitation
For Use With (Equipment) qPCR Instruments
Gene ID (Entrez) 3562
Gene Symbol IL-3; MCGF; MULTI-CSF
Incubation Time 1 hr.
Kit Contents Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Label or Dye FAM™
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Storage Requirements Frozen at -20°C
Target IL-3
Test Time 30 mins
Total Assay Time 2 hrs
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.