missing translation for 'onlineSavingsMsg'
Learn More

IL-3 Antibody (MP2-8F8), DyLight 650, Novus Biologicals™

Product Code. 18415653
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415653 0.5 mL 0.50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415653 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP151826C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-3 Monoclonal antibody specifically detects IL-3 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-3
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone MP2-8F8
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Hematopoietic growth factor, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Host Species Rat
Immunogen COS-expressed mouse IL-3
Purification Method Protein A or G purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3562
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.