missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-29 Mouse anti-Human, Clone: 1H3A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66163-1-IG-150UL
This item is not returnable.
View return policy
Description
IL29 is a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28A (IL28A), and interleukin 28B (IL28B) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).Specifications
| IL-29 | |
| Monoclonal | |
| 1.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IU54 | |
| IFNL1 | |
| IL-29 Fusion Protein Ag12444 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1H3A3 | |
| Unconjugated | |
| IFNL1 | |
| Cytokine Zcyto21, IFN lambda 1, IFNL1, IL 29, IL29, Interferon lambda 1, Interleukin 29, ZCYTO21 | |
| Mouse | |
| Protein G | |
| RUO | |
| 282618 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction