missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-29/IFN-lambda 1 Antibody (B-Z29) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18107JF585
This item is not returnable.
View return policy
Description
IL-29/IFN-lambda 1 Monoclonal antibody specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for ELISA
Specifications
| IL-29/IFN-lambda 1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| cytokine Zcyto21, IFNL1, IFN-lambda-1, IL-29, interferon lambda-1, interferon, lambda 1, interleukin 29 (interferon, lambda 1), interleukin-29, ZCYTO21 | |
| Recombinant human IL-29/IFN-lambda 1. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 282618 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| ELISA | |
| B-Z29 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction