missing translation for 'onlineSavingsMsg'
Learn More

IL-28R alpha/IFN-lambda R1 Antibody [DyLight 488], Novus Biologicals™

Product Code. 30737209 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737209 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737209 Supplier Novus Biologicals Supplier No. AF5260K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IL-28R alpha/IFN-lambda R1 Polyclonal antibody specifically detects IL-28R alpha/IFN-lambda R1 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-28R alpha/IFN-lambda R1
Applications Flow Cytometry, Western Blot, CyTOF
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias class II cytokine receptor CRF2/12, CRF2/12, CRF2-12, Cytokine receptor class-II member 12, Cytokine receptor family 2 member 12, IFN-lambda receptor 1, IFN-lambda-R1, IFNLR, IFNLR1, IL-28 receptor subunit alpha, IL-28R1, IL-28RA, IL-28R-alpha, Interferon lambda receptor 1, interferon lambda, receptor 1, interleukin 28 receptor A, interleukin 28 receptor, alpha, interleukin 28 receptor, alpha (interferon, lambda receptor), interleukin or cytokine receptor 2, interleukin-28 receptor subunit alpha, LICR2, Likely interleukin or cytokine receptor 2
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-28 R alpha/IFN-lambda R1, Arg21-Ala228, Accession # Q8IU57
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 163702
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.