missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-28R alpha/IFN-lambda R1 Antibody (601106) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5260JF669
This item is not returnable.
View return policy
Description
IL-28R alpha/IFN-lambda R1 Monoclonal antibody specifically detects IL-28R alpha/IFN-lambda R1 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| IL-28R alpha/IFN-lambda R1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| class II cytokine receptor CRF2/12, CRF2/12, CRF2-12, Cytokine receptor class-II member 12, Cytokine receptor family 2 member 12, IFN-lambda receptor 1, IFN-lambda-R1, IFNLR, IFNLR1, IL-28 receptor subunit alpha, IL-28R1, IL-28RA, IL-28R-alpha, Interferon lambda receptor 1, interferon lambda, receptor 1, interleukin 28 receptor A, interleukin 28 receptor, alpha, interleukin 28 receptor, alpha (interferon, lambda receptor), interleukin or cytokine receptor 2, interleukin-28 receptor subunit alpha, LICR2, Likely interleukin or cytokine receptor 2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-28 R alpha /IFN-lambda R1, Arg21-Ala228, Accession # Q8IU57 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 601106 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Cytokine Research | |
| 163702 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction