missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-28B/IFN-lambda 3 Antibody (120), DyLight 405 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90012V
This item is not returnable.
View return policy
Description
IL-28B/IFN-lambda 3 Monoclonal antibody specifically detects IL-28B/IFN-lambda 3 in Human samples. It is validated for ELISA
Specifications
| IL-28B/IFN-lambda 3 | |
| Monoclonal | |
| DyLight 405 | |
| interleukin-28B, IL28B, IL-28B, IL28C, interferon, lambda 3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-28B/IFN-lambda 3 (NP_742151.2; Met1-Val196). | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 282617 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 120 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction