Learn More
Invitrogen™ IL-28A/IL-28B Monoclonal Antibody (244716)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA523902
Description
In sandwich ELISAs, 100% cross-reactivity with recombinant mouse IL-28A/IFN-lambda 2 and no cross-reactivity with recombinant human (rh) IL-28A or rhIL-29 is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL28 is a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28B (IL28B), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).
Specifications
| IL-28A/IL-28B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q4VK74, Q8CGK6 | |
| Ifnl2, Ifnl3 | |
| E. coli-derived recombinant mouse IL-28B/IFN-lambda3 Asp20-Val193. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| ELISA, Neutralization | |
| 244716 | |
| Unconjugated | |
| Ifnl3 | |
| Cytokine Zcyto20; cytokine Zcyto22; EG330496; IFL-1; IFN lambda2; Ifnl2; Ifnl3; IFN-lambda2; IFN-lambda-2; IFN-lambda-3; IFN-lambda-4; IL 28; IL 28 A; Il28; Il28a; IL-28A; Il28b; IL-28B; IL28C; IL-28C; ILN; INF-alpha; INF-lambda; interferon alpha; interferon lambda 2; interferon lambda 3; Interferon lambda2; interferon lambda-2; Interferon lambda-3; interferon, lambda 2; interferon, lambda 3; interferon, lambda 4; interferon-lambda 2; interferon-lambda2; Interleukin; interleukin 28; interleukin 28A; interleukin 28A (interferon, lambda 2); interleukin 28B; interleukin-28A; Interleukin-28B; interleukin-28C; ZCYTO20; ZCYTO22 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 330496, 338374 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.