missing translation for 'onlineSavingsMsg'
Learn More

IL-28A/IFN-lambda 2 Antibody (248512) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30734919 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734919 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734919 Supplier Novus Biologicals Supplier No. FAB15871J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-28A/IFN-lambda 2 Monoclonal antibody specifically detects IL-28A/IFN-lambda 2 in Human samples. It is validated for ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-28A/IFN-lambda 2
Applications ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 248512
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias interleukin-28A, IL28A, IL-28A, interferon, lambda 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-28A/IFN-lambda 2, Val26-Val200, Accession # Q8IZJ0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 282616
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.