missing translation for 'onlineSavingsMsg'
Learn More

IL-27R alpha/WSX-1/TCCR Antibody (34N4G11), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18431516 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431516 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431516 Supplier Novus Biologicals Supplier No. NBP219015JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-27R alpha/WSX-1/TCCR Monoclonal antibody specifically detects IL-27R alpha/WSX-1/TCCR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow (Cell Surface), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-27R alpha/WSX-1/TCCR
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Cell Surface), CyTOF
Classification Monoclonal
Clone 34N4G11
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL27R, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1
Host Species Mouse
Immunogen A portion of amino acids 200-250 of human IL-27R (WSX-1) was used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 9466
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.