missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27 p28/IL-30 Antibody (234205), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1834P
This item is not returnable.
View return policy
Description
IL-27 p28/IL-30 Monoclonal antibody specifically detects IL-27 p28/IL-30 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| IL-27 p28/IL-30 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 234205 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| IL27,IL-27,IL27A,IL-27A,IL27p28,IL-27p28,IL30,p28 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse IL-27, Tyr19-Pro228 (EBI3) and Phe29-Ser234 (p28), Accession # O35228 (EBI3) and Q8K3I6 (p28) | |
| 0.5 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction