Learn More
Invitrogen™ IL-27 Monoclonal Antibody (355028)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA524101
Description
In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-27 p28, rmIL-27 EBI-3, or recombinant human (rh) IL-27 heterodimer is observed. In Western blots, no cross-reactivity with rmIL-27 p28 or rhIL-27 heterodimer is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
IL-27 (Interleukin 27) is a cytokine belonging to the IL-12 family, and is a heterodimeric cytokine composed of EBI3 and p28 subunits. IL-27 is produced by activated dendritic cells and macrophages in response to Toll-like receptor ligands and pro-inflammatory cytokines. IL-27 promotes CD4+ T cell differentiation to the Th1 lineage by inducing expression of the transcription factor T-bet and up regulating IL-12R beta2, which in turn suppresses Th2 and Th17 differentiation and proliferation. IL-27 is a recently discovered member of the IL-6/IL-12 family of proinflammatory and immunoregulatory cytokines. IL-27 acts by binding to its receptor WSX-1 and gp130 which results in the activation of a Jak/STAT signaling cascade, suggesting IL-27 involvement in the regulation of immune processes. It has been suggested that IL-27 can also be used as a therapeutic agent against cancer as it can also induce tumor-specific anti-tumor activity mediated through CD8+ T-cells, IFN-gamma, and T-bet. In humans, the gene is located on the p arm of chromosome 16.
Specifications
| IL-27 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O35228, Q8K3I6 | |
| Ebi3, IL27 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-27 heterodimer Tyr19-Pro228 (EBI3) and Phe29-Ser234 (p28). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunoprecipitation | |
| 355028 | |
| Unconjugated | |
| Ebi3 | |
| cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; Il27; IL-27; IL27 p28; IL-27 p28 subunit; IL27 subunit; IL-27 subunit alpha; IL-27 subunit beta; Il27a; IL-27A; IL-27-A; IL27-A; Il27b; IL-27B; IL27p28; IL-27p28; Il30; IL35 subunit; ILN; Interleukin; interleukin 27; interleukin 30; interleukin-27 subunit alpha; Interleukin-27 subunit beta; interleukin-30; MGC71873; M-IL-27 (p28); p28; RGD1561420 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 246779, 50498 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.