missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27 Monoclonal Antibody (355025), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524100
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse (rm) IL-27 p28, rmIL-27 EBI3, recombinant human (rh) IL-27 p28, and rhIL-27 EBI3 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
IL-27 Monoclonal antibody specifically detects IL-27 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IL-27 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O35228, Q8K3I6 | |
| Ebi3, IL27 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-27 heterodimer Tyr19-Pro228 (EBI3), Phe29-Ser234 (p28) | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 355025 | |
| Unconjugated | |
| IL27 | |
| cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; Il27; IL-27; IL27 p28; IL-27 p28 subunit; IL27 subunit; IL-27 subunit alpha; IL-27 subunit beta; Il27a; IL-27A; IL-27-A; IL27-A; Il27b; IL-27B; IL27p28; IL-27p28; Il30; IL35 subunit; ILN; Interleukin; interleukin 27; interleukin 30; interleukin-27 subunit alpha; Interleukin-27 subunit beta; interleukin-30; MGC71873; M-IL-27 (p28); p28; RGD1561420 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 246779, 50498 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction