missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76698-0.025mg
This item is not returnable.
View return policy
Description
IL-27 Polyclonal specifically detects IL-27 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry.
Specifications
| IL-27 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A | |
| Rabbit | |
| 28 kDa | |
| 0.025 mg | |
| Cell Cycle and Replication | |
| 246778 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-4 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
| NP_663634 | |
| IL27 | |
| Antibody was raised against a peptide corresponding to 16 amino acids near the amino terminus of human IL-27. The immunogen is located within the first 50 amino acids of IL-27. Amino Acid Squence: WGFPRPPGRPQLSLQE | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human IL-27 has one isoform (243aa, 28kD). Mouse IL-27 has one isoform (234aa, 27kD) and rat IL-27 also has one isoform (234aa, 27kD). NBP1-76698 can detect human, mouse and rat IL-27. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction