missing translation for 'onlineSavingsMsg'
Learn More

IL-27 Antibody (355028) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30732791 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732791 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732791 Supplier Novus Biologicals Supplier No. FAB18343JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-27 Monoclonal antibody specifically detects IL-27 in Mouse samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-27
Applications Immunoprecipitation
Classification Monoclonal
Clone 355028
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-27 heterodimer, Tyr19-Pro228 (EBI3) and Phe29-Ser234 (p28), Accession # O35228 (EBI3) & Q8K3I6 (p28)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 246778
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.