missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-26 Monoclonal Antibody (510414), APC
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523643
This item is not returnable.
View return policy
Description
IL-26 Monoclonal Antibody for Flow
IL26 is a member of the IL10 family of cytokines. It is a secreted protein and may function as a homodimer. This protein is thought to contribute to the transformed phenotype of T cells after infection by herpesvirus samimiri.
Specifications
| IL-26 | |
| Monoclonal | |
| APC | |
| IL26 | |
| AK155; il 26; IL26; IL-26; ILN; Interleukin; interleukin 26; interleukin-26; Protein AK155 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 55801 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 510414 | |
| Buffered saline solution with ≤0.5% BSA and 0.09% sodium azide | |
| Q9NPH9 | |
| IL26 | |
| E. coli-derived recombinant human IL-26/AK155 Lys22-Gln171. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction