missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-26/AK155 Antibody (MM0399-6D20), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11703G
This item is not returnable.
View return policy
Description
IL-26/AK155 Monoclonal specifically detects IL-26/AK155 in Human samples. It is validated for Western Blot.
Specifications
| IL-26/AK155 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IL26 | |
| Human recombinant IL-26/AK155 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0399-6D20 | |
| Western Blot | |
| AK155IL-26interleukin-26, interleukin 26, Protein AK155 | |
| Mouse | |
| Protein G purified | |
| Cytokine Research | |
| 55801 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction