missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-26/AK155 Antibody (510414) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB13751E
This item is not returnable.
View return policy
Description
IL-26/AK155 Monoclonal antibody specifically detects IL-26/AK155 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| IL-26/AK155 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 510414 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| AK155IL-26interleukin-26, interleukin 26, Protein AK155 | |
| E. coli-derived recombinant human IL-26/AK155, Lys22-Gln171, Accession # Q9NPH9.1 | |
| 0.1 mL | |
| Cytokine Research | |
| 55801 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction