missing translation for 'onlineSavingsMsg'
Learn More

IL-26/AK155 Antibody (197505) [DyLight 594], Novus Biologicals™

Product Code. 30733278 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733278 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733278 Supplier Novus Biologicals Supplier No. FAB1375M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-26/AK155 Monoclonal antibody specifically detects IL-26/AK155 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-26/AK155
Applications Western Blot
Classification Monoclonal
Clone 197505
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias AK155IL-26interleukin-26, interleukin 26, Protein AK155
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-26, Lys22-Gln171, Accession # Q9NPH9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 55801
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.