missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-25 Monoclonal Antibody (182203), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523568
This item is not returnable.
View return policy
Description
This antibody shows 100% cross-reactivity with recombinant mouse IL-17E and no cross-reactivity with recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, or rhIL-17F.
Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
Specifications
| IL-25 | |
| Monoclonal | |
| PE | |
| IL25 | |
| D17Wsu104e; il 17e; Il17e; IL-17E; Il25; IL-25; ILN; Interleukin; interleukin 17E; interleukin 25; interleukin-17E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homolog | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 64806 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 182203 | |
| Buffered saline solution with BSA and 0.09% sodium azide | |
| Q9H293 | |
| IL25 | |
| E. coli-derived recombinant human IL-17E Tyr33-Gly177. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction