missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-25/IL-17E ELISA Kit

Product Code. 15582007
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15582007 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15582007 Supplier Invitrogen™ Supplier No. BMS6046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Mouse Interleukin-25/Interleukin 17E (Ms IL-25/IL-17E) ELISA quantitates Ms IL-25/IL-17E in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-25/IL-17E. Principle of the method The Mouse IL-25/IL-17E solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8VHH8
Assay Range 3.9-250 pg/mL
Assay Sensitivity 0.9 pg/mL
Content And Storage Aluminum pouch(es) with a Microplate coated with monoclonal antibody to mouse IL-17E (IL-25)
Biotin-Conjugate anti-mouse IL-17E (IL-25) monoclonal antibody
Streptavidin-HRP
Mouse IL-17E (IL-25) Standard lyophilized, 500 pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias D17Wsu104e; il 17e; Il17e; IL-17E; Il25; IL-25; ILN; Interleukin; interleukin 17E; interleukin 25; interleukin-17E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homologD17Wsu104e; il 17e; Il17e; IL-17E; Il25; IL-25; ILN; Interleukin; interleukin 17E; interleukin 25; interleukin-17E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homolog; IL-17E; Il17e
Gene ID (Entrez) 140806
Gene Symbol IL25
Interassay CV 8.8%
Intraassay CV 6.5%
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2-8°C
Synonym Interleukin-17 E, Interleukin-25
Target IL-25
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.