missing translation for 'onlineSavingsMsg'
Learn More

IL-24 Antibody (MM0398-6F48), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18432896 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432896 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432896 Supplier Novus Biologicals Supplier No. NBP211702JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-24 Monoclonal antibody specifically detects IL-24 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-24
Applications Western Blot
Classification Monoclonal
Clone MM0398-6F48
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias C49A, FISP, IL10B, IL-24, interleukin 24, mda-7, MDA7MOB5, melanocyte-associated Mda-7, melanoma differentiation association protein 7, Melanoma differentiation-associated gene 7 protein, Mob-5, ST16interleukin-24, suppression of tumorigenicity 16 (melanoma differentiation), Suppression of tumorigenicity 16 protein
Host Species Mouse
Immunogen Human recombinant IL-24
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 11009
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.