missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-23A/IL-23 P19 Antibody (727753) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB17161JF525
This item is not returnable.
View return policy
Description
IL-23A/IL-23 P19 Monoclonal antibody specifically detects IL-23A/IL-23 P19 in Human samples. It is validated for ELISA,Western Blot
Specifications
| IL-23A/IL-23 P19 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| IL-23, IL-23A, IL-23-A, IL-23p19, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRFIL-23 subunit alpha | |
| E. coli-derived recombinant human IL-23 p19, Arg20-Pro189, Accession # Q9NPF7 | |
| 0.1 mL | |
| Cancer, Cytokine Research, Innate Immunity | |
| 51561 | |
| Store at 4°C in the dark. | |
| IgG2b |
| ELISA, Western Blot | |
| 727753 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction