missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-23A/IL-23 P19 Monoclonal antibody specifically detects IL-23A/IL-23 P19 in Human samples. It is validated for Western Blot,Neutralization
Specifications
Specifications
| Antigen | IL-23A/IL-23 P19 |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Clone | 1193B |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | IL-23, IL-23A, IL-23-A, IL-23p19, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRFIL-23 subunit alpha |
| Host Species | Rabbit |
| Immunogen | E. coli-derived recombinant human IL-23 p19, Arg20-Pro189, Accession # Q9NPF7 |
| Purification Method | Protein A or G purified from cell culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?